Search
Search

Kpna6| Gene Symbol: Kpna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500641.2 | XP_003500689.1 | 9 - 10 | 986 | 60 | |
| XM_007641405.1 | XP_007639595.1 | 9 - 10 | 985 | 60 | ||
| GenBank mRNA | JI888406.1 | - | 9 - 10 | 978 | 60 | |
| JP062685.1 | - | 9 - 10 | 978 | 60 | ||
Set Membership: |
|