Search
Search

LOC100767530| Gene Symbol: LOC100767530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514944.2 | XP_003514992.1 | 3 - 4 | 226 | 83 | |
| XM_007654969.1 | XP_007653159.1 | 3 - 4 | 227 | 83 | ||
Set Membership: |
|