Search
Search
| Gene Symbol: TNIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675313.3 | XP_005675370.1 | 31 - 32 | 4614 | 81 | |
| XM_005675314.3 | XP_005675371.1 | 30 - 31 | 4589 | 81 | ||
| XM_005675315.3 | XP_005675372.1 | 30 - 31 | 4526 | 81 | ||
| XM_005675316.3 | XP_005675373.1 | 29 - 30 | 4502 | 81 | ||
| XM_013964598.2 | XP_013820052.1 | 32 - 33 | 4701 | 81 | ||
| XM_013964609.2 | XP_013820063.1 | 31 - 32 | 4614 | 81 | ||
Set Membership: |
|