Search
Search
| Gene Symbol: WNK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065141.1 | XP_017920630.1 | 1 - 2 | 733 | 80 | |
| XM_018065142.1 | XP_017920631.1 | 1 - 2 | 733 | 80 | ||
| XM_018065143.1 | XP_017920632.1 | 1 - 2 | 733 | 80 | ||
| XM_018065145.1 | XP_017920634.1 | 2 - 3 | 975 | 80 | ||
Set Membership: |
|