Search
Search

CEP170| Gene Symbol: CEP170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060262.1 | XP_017915751.1 | 13 - 14 | 4818 | 64 | |
| XM_018060263.1 | XP_017915752.1 | 13 - 14 | 4818 | 64 | ||
| XM_018060264.1 | XP_017915753.1 | 13 - 14 | 4818 | 64 | ||
| XM_018060265.1 | XP_017915754.1 | 13 - 14 | 4818 | 64 | ||
| XM_018060266.1 | XP_017915755.1 | 13 - 14 | 4710 | 64 | ||
| XM_018060267.1 | XP_017915756.1 | 13 - 14 | 4818 | 64 | ||
| XM_018060268.1 | XP_017915757.1 | 14 - 15 | 4630 | 64 | ||
| XM_018060269.1 | XP_017915758.1 | 14 - 15 | 4630 | 64 | ||
| XM_018060271.1 | XP_017915760.1 | 13 - 14 | 4336 | 64 | ||
| XM_018060272.1 | XP_017915761.1 | 13 - 14 | 4336 | 64 | ||
| XM_018060273.1 | XP_017915762.1 | 13 - 14 | 4229 | 64 | ||
Set Membership: |
|