Search
Search
| Gene Symbol: DTNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055540.1 | XP_017911029.1 | 2 - 3 | 303 | 81 | |
| XM_018055542.1 | XP_017911031.1 | 2 - 3 | 303 | 81 | ||
| XM_018055543.1 | XP_017911032.1 | 2 - 3 | 303 | 81 | ||
| XM_018055544.1 | XP_017911033.1 | 2 - 3 | 303 | 81 | ||
| XM_018055545.1 | XP_017911034.1 | 2 - 3 | 303 | 81 | ||
| XM_018055546.1 | XP_017911035.1 | 2 - 3 | 303 | 81 | ||
| GenBank mRNA | JO442709.2 | - | 2 - 3 | 307 | 81 | |
| JO593528.2 | - | 2 - 3 | 185 | 81 | ||
Set Membership: |
|