Search
Search

LOC102169209| Gene Symbol: LOC102169209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060792.1 | XP_017916281.1 | 7 - 8 | 1176 | 90 | |
| XM_018060793.1 | XP_017916282.1 | 7 - 8 | 1176 | 90 | ||
| XM_018060795.1 | XP_017916284.1 | 7 - 8 | 1176 | 90 | ||
| XM_018060796.1 | XP_017916285.1 | 7 - 8 | 1176 | 90 | ||
| XM_018060797.1 | XP_017916286.1 | 7 - 8 | 1176 | 90 | ||
Set Membership: |
|