Search
Search
| Gene Symbol: NRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057070.1 | XP_017912559.1 | 4 - 5 | 1170 | 60 | |
| XM_018057071.1 | XP_017912560.1 | 4 - 5 | 1170 | 60 | ||
| XM_018057072.1 | XP_017912561.1 | 4 - 5 | 1170 | 60 | ||
| XM_018057073.1 | XP_017912562.1 | 4 - 5 | 1170 | 60 | ||
| GenBank mRNA | JO425394.1 | - | 4 - 5 | 1018 | 60 | |
Set Membership: |
|