Search
Search

ELMSAN1| Gene Symbol: ELMSAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053988.1 | XP_017909477.1 | 12 - 13 | 3931 | 71 | |
| XM_018053989.1 | XP_017909478.1 | 12 - 13 | 4329 | 71 | ||
Set Membership: |
|