Search
Search
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699038.3 | XP_005699095.1 | 6 - 7 | 340 | 66 | |
| XM_013975553.2 | XP_013831007.1 | 6 - 7 | 316 | 66 | ||
| GenBank mRNA | JO458816.2 | - | 6 - 7 | 304 | 66 | |
Set Membership: |
|