Search
Search
| Gene Symbol: CD226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696957.3 | XP_005697014.2 | 3 - 4 | 1085 | 66 | |
| GenBank mRNA | JO322829.2 | - | 3 - 4 | 715 | 66 | |
| JO322831.2 | - | 3 - 4 | 715 | 66 | ||
Set Membership: |
|