Search
Search

SH2D3C| Gene Symbol: SH2D3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687163.3 | XP_005687220.2 | 11 - 12 | 2525 | 81 | |
| XM_005687164.3 | XP_005687221.2 | 10 - 11 | 1992 | 81 | ||
| XM_018055821.1 | XP_017911310.1 | 9 - 10 | 2000 | 81 | ||
| GenBank mRNA | JO333744.2 | - | 7 - 8 | 1401 | 81 | |
Set Membership: |
|