Search
Search

SLC22A16| Gene Symbol: SLC22A16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966433.2 | XP_013821887.2 | 6 - 7 | 2273 | 76 | |
| XM_018053071.1 | XP_017908560.1 | 7 - 8 | 1607 | 76 | ||
Set Membership: |
|