Search
Search
| Gene Symbol: TDRD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052198.1 | XP_017907687.1 | 16 - 17 | 3641 | 60 | |
| XM_018052199.1 | XP_017907688.1 | 15 - 16 | 3109 | 60 | ||
| GenBank mRNA | JT636313.1 | - | 16 - 17 | 3057 | 60 | |
Set Membership: |
|