Search
Search
| Gene Symbol: ITGA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064673.1 | XP_017920162.1 | 12 - 13 | 1916 | 61 | |
| XM_018064676.1 | XP_017920165.1 | 12 - 13 | 1915 | 61 | ||
| GenBank mRNA | JR128439.1 | - | 155 | 61 | ||
Set Membership: |
|