Search
Search
| Gene Symbol: GKAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052412.1 | XP_017907901.1 | 9 - 10 | 1329 | 82 | |
| XM_018052413.1 | XP_017907902.1 | 9 - 10 | 1120 | 82 | ||
| XM_018052414.1 | XP_017907903.1 | 9 - 10 | 1030 | 82 | ||
| XM_018052415.1 | XP_017907904.1 | 8 - 9 | 1176 | 82 | ||
| GenBank mRNA | KA313999.1 | - | 4 - 5 | 539 | 82 | |
Set Membership: |
|