Search
Search
| Gene Symbol: ARMH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041317.1 | XP_017896806.1 | 9 - 10 | 950 | 94 | |
| XM_018041318.1 | XP_017896807.1 | 8 - 9 | 813 | 94 | ||
| XM_018041319.1 | XP_017896808.1 | 9 - 10 | 878 | 94 | ||
Set Membership: |
|