Search
Search

ITCH| Gene Symbol: ITCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747264.2 | XP_002747310.1 | 14 - 15 | 1638 | 69 | |
| XM_008995498.1 | XP_008993746.1 | 9 - 10 | 1056 | 69 | ||
| XM_017971823.1 | XP_017827312.1 | 4 - 5 | 363 | 69 | ||
Set Membership: |
|