Search
Search

EHMT2| Gene Symbol: EHMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994184.2 | XP_008992432.1 | 22 - 23 | 2887 | 75 | |
| XM_017970980.1 | XP_017826469.1 | 21 - 22 | 2785 | 75 | ||
Set Membership: |
|