Search
Search

ATP11B| Gene Symbol: ATP11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981276.2 | XP_008979524.1 | 20 - 21 | 7276 | 82 | |
| XM_017964706.1 | XP_017820195.1 | 20 - 21 | 7280 | 82 | ||
| XM_017964707.1 | XP_017820196.1 | 20 - 21 | 7280 | 82 | ||
| XM_017964708.1 | XP_017820197.1 | 20 - 21 | 7280 | 82 | ||
| XM_017964709.1 | XP_017820198.1 | 20 - 21 | 7280 | 82 | ||
Set Membership: |
|