Search
Search

LRRK2| Gene Symbol: LRRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752367.3 | XP_002752413.1 | 33 - 34 | 4944 | 63 | |
| XM_009003633.2 | XP_009001881.1 | 34 - 35 | 5254 | 63 | ||
| XM_009003634.2 | XP_009001882.1 | 34 - 35 | 5256 | 63 | ||
| XM_009003635.2 | XP_009001883.1 | 33 - 34 | 5120 | 63 | ||
| XM_017975992.1 | XP_017831481.1 | 32 - 33 | 4615 | 63 | ||
Set Membership: |
|