Search
Search

USP49| Gene Symbol: USP49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746532.4 | XP_002746578.1 | 1 - 2 | 1356 | 109 | |
| XM_008994421.2 | XP_008992669.1 | 1 - 2 | 1356 | 109 | ||
Set Membership: |
|