Search
Search

DYRK1A| Gene Symbol: DYRK1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761413.4 | XP_002761459.1 | 7 - 8 | 5607 | 84 | |
| XM_008986673.2 | XP_008984921.1 | 7 - 8 | 2522 | 84 | ||
Set Membership: |
|