Search
Search

LTBP1| Gene Symbol: LTBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980891.2 | XP_008979139.1 | 9 - 10 | 2294 | 82 | |
| XM_008980892.2 | XP_008979140.1 | 9 - 10 | 2294 | 82 | ||
| XM_008980893.2 | XP_008979141.1 | 9 - 10 | 2294 | 82 | ||
| XM_008980894.2 | XP_008979142.1 | 9 - 10 | 2294 | 82 | ||
| XM_008980895.2 | XP_008979143.1 | 9 - 10 | 2294 | 82 | ||
| XM_008980897.2 | XP_008979145.1 | 5 - 6 | 1445 | 82 | ||
| XM_008980898.2 | XP_008979146.1 | 5 - 6 | 1434 | 82 | ||
| XM_008980899.2 | XP_008979147.1 | 5 - 6 | 1434 | 82 | ||
| XM_017964425.1 | XP_017819914.1 | 7 - 8 | 1507 | 82 | ||
| XM_017964426.1 | XP_017819915.1 | 7 - 8 | 2290 | 82 | ||
Set Membership: |
|