Search
Search

TANGO6| Gene Symbol: TANGO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986128.2 | XP_008984376.1 | 8 - 9 | 1583 | 106 | |
| XM_017966800.1 | XP_017822289.1 | 8 - 9 | 1583 | 106 | ||
| XM_017966801.1 | XP_017822290.1 | 8 - 9 | 1583 | 106 | ||
| XM_017966802.1 | XP_017822291.1 | 8 - 9 | 1583 | 106 | ||
Set Membership: |
|