Search
Search

ITFG2| Gene Symbol: ITFG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975863.1 | XP_017831352.1 | 9 - 10 | 995 | 63 | |
| XM_017975864.1 | XP_017831353.1 | 9 - 10 | 995 | 63 | ||
| XM_017975865.1 | XP_017831354.1 | 7 - 8 | 857 | 63 | ||
Set Membership: |
|