Search
Search

CD44| Gene Symbol: CD44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755175.3 | XP_002755221.1 | 17 - 18 | 2424 | 72 | |
| XM_002755176.3 | XP_002755222.1 | 16 - 17 | 2295 | 72 | ||
| XM_009007913.1 | XP_009006161.1 | 16 - 17 | 2307 | 72 | ||
| XM_009007915.1 | XP_009006163.1 | 15 - 16 | 2178 | 72 | ||
| XM_009007916.1 | XP_009006164.1 | 15 - 16 | 2178 | 72 | ||
| XM_009007917.1 | XP_009006165.1 | 15 - 16 | 2178 | 72 | ||
| XM_009007918.1 | XP_009006166.1 | 15 - 16 | 2103 | 72 | ||
| XM_009007919.1 | XP_009006167.1 | 14 - 15 | 2052 | 72 | ||
| XM_009007920.1 | XP_009006168.1 | 14 - 15 | 2049 | 72 | ||
| XM_009007921.1 | XP_009006169.1 | 13 - 14 | 1935 | 72 | ||
| XM_009007922.1 | XP_009006170.1 | 12 - 13 | 1806 | 72 | ||
| XM_009007925.1 | XP_009006173.1 | 11 - 12 | 1680 | 72 | ||
| XM_009007926.1 | XP_009006174.1 | 9 - 10 | 1488 | 72 | ||
| XM_009007927.1 | XP_009006175.1 | 10 - 11 | 1476 | 72 | ||
| XM_009007928.1 | XP_009006176.1 | 8 - 9 | 1284 | 72 | ||
| XM_009007931.1 | XP_009006179.1 | 3 - 4 | 496 | 72 | ||
| GenBank mRNA | EF613222.1 | - | 8 - 9 | 1001 | 72 | |
Set Membership: |
|