Search
Search

SKA2| Gene Symbol: SKA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748165.4 | XP_002748211.1 | 2 - 3 | 227 | 102 | |
| XM_008996946.2 | XP_008995194.1 | 2 - 3 | 173 | 102 | ||
| XM_008996947.2 | XP_008995195.1 | 2 - 3 | 192 | 102 | ||
| XM_017972479.1 | XP_017827968.1 | 2 - 3 | 121 | 102 | ||
Set Membership: |
|