Search
Search

WDR66| Gene Symbol: WDR66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753109.4 | XP_002753155.2 | 2 - 3 | 805 | 93 | |
| XM_009004808.2 | XP_009003056.1 | 2 - 3 | 561 | 93 | ||
| XM_009004809.2 | XP_009003057.1 | 2 - 3 | 913 | 93 | ||
| XM_009004812.2 | XP_009003060.1 | 2 - 3 | 805 | 93 | ||
| XM_009004814.2 | XP_009003062.1 | 2 - 3 | 805 | 93 | ||
| XM_017976446.1 | XP_017831935.1 | 2 - 3 | 805 | 93 | ||
| XM_017976448.1 | XP_017831937.1 | 2 - 3 | 805 | 93 | ||
Set Membership: |
|