Search
Search

ROCK1| Gene Symbol: ROCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979587.2 | XP_008977835.1 | 33 - 34 | 4133 | 76 | |
| XM_017963914.1 | XP_017819403.1 | 32 - 33 | 3910 | 76 | ||
Set Membership: |
|