Search
Search

ZNF333| Gene Symbol: ZNF333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987419.2 | XP_008985667.1 | 5 - 6 | 594 | 69 | |
| XM_008987420.2 | XP_008985668.1 | 5 - 6 | 399 | 69 | ||
| XM_008987421.2 | XP_008985669.1 | 5 - 6 | 594 | 69 | ||
| XM_008987423.2 | XP_008985671.1 | 4 - 5 | 469 | 69 | ||
Set Membership: |
|