Search
Search

MOBP| Gene Symbol: MOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965965.1 | XP_017821454.1 | 3 | 325 | 85 | |
| XM_017965966.1 | XP_017821455.1 | 3 | 324 | 85 | ||
| XM_017965967.1 | XP_017821456.1 | 3 | 325 | 85 | ||
| XM_017965968.1 | XP_017821457.1 | 3 | 317 | 85 | ||
Set Membership: |
|