Search
Search

SERPINB13| Gene Symbol: SERPINB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979933.2 | XP_008978181.1 | 3 - 4 | 363 | 89 | |
| XM_017963626.1 | XP_017819115.1 | 3 - 4 | 381 | 89 | ||
Set Membership: |
|