Search
Search

SLAMF7| Gene Symbol: SLAMF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760177.3 | XP_002760223.1 | 1 - 2 | 770 | 66 | |
| XM_008984744.2 | XP_008982992.1 | 1 - 2 | 766 | 66 | ||
Set Membership: |
|