Search
Search

CG15270| Gene Symbol: CG15270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259105.1 | NP_001246034.1 | 4 - 5 | 2023 | 69 | |
| NM_001273530.1 | NP_001260459.1 | 5 - 6 | 1399 | 69 | ||
| NM_001299022.1 | NP_001285951.1 | 5 - 6 | 1399 | 69 | ||
| NM_001299023.1 | NP_001285952.1 | 5 - 6 | 1399 | 69 | ||
| NM_135894.3 | NP_609738.1 | 4 - 5 | 2023 | 69 | ||
| NM_205997.2 | NP_995719.1 | 4 - 5 | 2023 | 69 | ||
Set Membership: |
|