Search
Search

coro| Gene Symbol: coro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273813.1 | NP_001260742.1 | 7 - 8 | 1621 | 85 | |
| NM_001273814.1 | NP_001260743.1 | 6 - 7 | 1668 | 85 | ||
| NM_001273815.1 | NP_001260744.1 | 6 - 7 | 1507 | 85 | ||
| NM_001273816.1 | NP_001260745.1 | 6 - 7 | 1482 | 85 | ||
| NM_136398.4 | NP_610242.1 | 6 - 7 | 1603 | 85 | ||
| NM_165494.2 | NP_724509.1 | 6 - 7 | 1503 | 85 | ||
| NM_206031.2 | NP_995753.1 | 6 - 7 | 1582 | 85 | ||
| GenBank mRNA | AF467271.1 | - | 6 - 7 | 1564 | 85 | |
| AY118560.1 | - | 6 - 7 | 1654 | 85 | ||
Set Membership: |
|