Search
Search

HGTX| Gene Symbol: HGTX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382022.1 | NP_001368954.1 | 2 - 3 | 1942 | 56 | |
| NM_144357.3 | NP_652614.1 | 2 - 3 | 1942 | 56 | ||
| GenBank mRNA | AF220236.2 | - | 2 - 3 | 1970 | 56 | |
| BT015992.1 | - | 2 - 3 | 1939 | 56 | ||
| BT022198.1 | - | 2 - 3 | 2147 | 56 | ||
Set Membership: |
|