Search
Search

olf413| Gene Symbol: olf413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275275.1 | NP_001262204.1 | 6 - 7 | 2363 | 61 | |
| NM_168948.5 | NP_730711.3 | 6 - 7 | 2363 | 61 | ||
| GenBank mRNA | BT011070.1 | - | 6 - 7 | 2337 | 61 | |
Set Membership: |
|