Search
Search

Tmc| Gene Symbol: Tmc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259769.3 | NP_001246698.1 | 16 - 17 | 4265 | 69 | |
| NM_001316433.1 | NP_001303362.1 | 17 - 18 | 4314 | 69 | ||
Set Membership: |
|