Search
Search

bsk| Gene Symbol: bsk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169459.1 | NP_001162930.1 | 2 - 3 | 780 | 91 | |
| NM_001169461.3 | NP_001162932.1 | 2 - 3 | 638 | 91 | ||
| NM_164900.3 | NP_723541.1 | 3 - 4 | 518 | 91 | ||
| GenBank mRNA | AY070865.1 | - | 3 - 4 | 395 | 91 | |
| AY122221.1 | - | 3 - 4 | 497 | 91 | ||
| U49180.1 | - | 2 - 3 | 781 | 91 | ||
Set Membership: |
|