Search
Search

Oscillin| Gene Symbol: Oscillin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258965.2 | NP_001245894.1 | 4 - 5 | 435 | 123 | |
| NM_001258966.1 | NP_001245895.1 | 3 - 4 | 445 | 123 | ||
| NM_001258967.2 | NP_001245896.1 | 4 - 5 | 476 | 123 | ||
| NM_001258968.1 | NP_001245897.1 | 3 - 4 | 445 | 123 | ||
| NM_001258969.2 | NP_001245898.1 | 3 - 4 | 702 | 123 | ||
| NM_001273172.1 | NP_001260101.1 | 3 - 4 | 445 | 123 | ||
| NM_135094.2 | NP_608938.1 | 3 - 4 | 445 | 123 | ||
| NM_164649.2 | NP_723092.1 | 2 - 3 | 753 | 123 | ||
| GenBank mRNA | AY122220.1 | - | 3 - 4 | 445 | 123 | |
Set Membership: |
|