Search
Search

Gr39a| Gene Symbol: Gr39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165370.1 | NP_724329.1 | 3 - 4 | 1033 | 110 | |
| NM_165371.1 | NP_724330.1 | 3 - 4 | 1060 | 110 | ||
| NM_165372.1 | NP_724331.1 | 3 - 4 | 1060 | 110 | ||
| NM_165373.1 | NP_724332.1 | 3 - 4 | 1030 | 110 | ||
Set Membership: |
|