Search
Search

wde| Gene Symbol: wde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299365.1 | NP_001286294.1 | 4 - 5 | 3898 | 64 | |
| NM_136764.3 | NP_610608.1 | 4 - 5 | 4047 | 64 | ||
| GenBank mRNA | AF145688.1 | - | 4 - 5 | 4034 | 64 | |
Set Membership: |
|