Search
Search

mirr| Gene Symbol: mirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274849.2 | NP_001261778.1 | 4 - 5 | 1624 | 123 | |
| NM_079323.4 | NP_524047.2 | 4 - 5 | 1624 | 123 | ||
| NM_168505.3 | NP_729818.1 | 4 - 5 | 1624 | 123 | ||
Set Membership: |
|