Search
Search

Deaf1| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038944.2 | NP_001034033.1 | 3 - 4 | 1911 | 58 | |
| NM_001275139.1 | NP_001262068.1 | 2 - 3 | 1537 | 58 | ||
| NM_001275140.1 | NP_001262069.1 | 3 - 4 | 1920 | 58 | ||
| NM_001275141.1 | NP_001262070.1 | 2 - 3 | 1528 | 58 | ||
| NM_079445.3 | NP_524169.1 | 3 - 4 | 1920 | 58 | ||
| GenBank mRNA | BT004907.1 | - | 3 - 4 | 1886 | 58 | |
| BT125910.1 | - | 2 - 3 | 1537 | 58 | ||
| U46686.1 | - | 3 - 4 | 1919 | 58 | ||
Set Membership: |
|