Search
Search

Lrp4| Gene Symbol: Lrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169298.2 | NP_001162769.1 | 5 - 6 | 1969 | 74 | |
| NM_001298378.1 | NP_001285307.1 | 5 - 6 | 2138 | 74 | ||
| NM_167474.3 | NP_727914.1 | 5 - 6 | 1969 | 74 | ||
| GenBank mRNA | BT046152.1 | - | 4 - 5 | 1859 | 74 | |
Set Membership: |
|