Search
Search

mRpS14| Gene Symbol: mRpS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298509.1 | NP_001285438.1 | 1 - 2 | 432 | 68 | |
| NM_167650.2 | NP_728245.1 | 1 - 2 | 111 | 68 | ||
| GenBank mRNA | BT023293.1 | - | 1 - 2 | 111 | 68 | |
| KX532149.1 | - | 1 - 2 | 70 | 68 | ||
Set Membership: |
|