Search
Search

frm| Gene Symbol: frm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104040.2 | NP_001097510.1 | 12 - 13 | 2141 | 60 | |
| NM_139713.5 | NP_647970.1 | 9 - 10 | 994 | 60 | ||
| NM_168119.4 | NP_729073.1 | 8 - 9 | 1210 | 60 | ||
| NM_168120.4 | NP_729074.1 | 11 - 12 | 2223 | 60 | ||
| NM_168121.4 | NP_729075.1 | 12 - 13 | 2264 | 60 | ||
| GenBank mRNA | AY069625.1 | - | 8 - 9 | 974 | 60 | |
| BT003763.1 | - | 12 - 13 | 2141 | 60 | ||
Set Membership: |
|