Search
Search

CG3776| Gene Symbol: CG3776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299939.1 | NP_001286868.1 | 2 - 3 | 452 | 85 | |
| NM_001299940.1 | NP_001286869.1 | 1 - 2 | 385 | 85 | ||
| NM_138145.3 | NP_611989.1 | 1 - 2 | 385 | 85 | ||
| GenBank mRNA | AY070543.1 | - | 1 - 2 | 369 | 85 | |
Set Membership: |
|