Search
Search

CG15369| Gene Symbol: CG15369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298112.1 | NP_001285041.1 | 1 - 2 | 222 | 56 | |
| NM_132314.3 | NP_572542.1 | 1 - 2 | 222 | 56 | ||
| GenBank mRNA | KX531447.1 | - | 1 - 2 | 168 | 56 | |
Set Membership: |
|